Buffers in Biology Lab Activity
Supplier: Avantor
470012-254EA 125.79 USD
470012-254
Buffers in Biology Lab Activity
Educational Classroom Kits and Activities
Discover what buffers exist in potatoes and liver.
Students explore the buffering capabilities of plant and animal tissues you provide to determine the buffers that exist naturally in various substances and solutions. Through the investigation, students learn that living things have buffers in them in order to stabilize their pH. This understanding can then be related to the effect of fluctuating pH on enzyme activity and relate acid/base chemistry to anatomy and physiology. Developed by Julie Bohrer, the activity includes instructions. A blender and fresh biomaterials are required, but not included.