Ward's® Chemistry Blue Bottle Reaction Demonstration
Supplier: Avantor
470011-896EA 65.89 USD
470011-896
Ward's® Chemistry Blue Bottle Reaction Demonstration
Educational Classroom Kits and Activities
Amaze Your Students with this Reversible Reaction.
  • Show how increasing the oxygen in the bottle, changes the color of the solution
  • Takes about 20 minutes to complete
Compounds that change color as a result of a change in their pH or oxidation state are called indicators. With this activity, you can illustrate this principle to your students by mixing a solution to introduce oxygen from the air into the solution. When you stop mixing, the reaction will reverse itself and become clear again. You will get instructions, the indicator solution, and two chemicals preweighed for a one liter demonstration. This demonstration can be completed in 20 minutes.
Ordering information: Set includes D-Glucose Monohydrate, Methylene Blue, Potassium Hydroxide, and Instructions. Required but not provided include 1 L bottles and General Labware.
Delivery information: Due to the nature of this item, it cannot be shipped by air under any circumstances. Please select ground shipping at time of checkout.