Ward's® Chemical Battery Demonstration
Supplier: Avantor
470010-848EA 70.19 USD
470010-848
Ward's® Chemical Battery Demonstration
Educational Classroom Kits and Activities
Construct a Simple Chemical Wet-Cell Battery.
  • The perfect supplement to your chemistry lectures
  • Can be completed in 15 minutes
  • Requires only basic labware that you provide
This easily-constructed battery can be built using two chemical solutions and two metal electrodes in an Erlenmeyer flask. It produces a constant source of 1.5 V and can be used to power a small LED. Using the materials included in this kit, you can show your students how to construct a single battery of this type.