Ward's® Chemistry Forming Red, White and Blue Demonstration
Supplier: Avantor
470010-844EA 64.89 USD
470010-844
Ward's® Chemistry Forming Red, White and Blue Demonstration
Educational Classroom Kits and Activities
A Great Way to Introduce Chemical Reaction Types
  • Provide for interesting discussion of 3 different chemical reactions
This “patriotic” demonstration illustrates complex ion formation, double replacement reaction and Prussian blue. Three reactions are carried out in adjacent beakers resulting in red, white and blue solutions.
Ordering information: Includes iron(III) chloride solution, potassium ferrocyanide solution, potassium thiocyanate solution, silver nitrate solution.