Ward's® Chemistry Acetylene-Chlorine Reaction Demonstration
Supplier: Avantor
470010-830EA 36.29 USD
470010-830
Ward's® Chemistry Acetylene-Chlorine Reaction Demonstration
Educational Classroom Kits and Activities
Study exothermic and oxidation-reduction reactions with ease.
  • Aligned to National Science Education standards
  • This demonstration can be performed eight times
This reaction demonstrates the reaction of acetylene and chlorine to form hydrochloric acid and carbon. This dramatic demonstration of a spontaneous reaction also generates light as a product.