Ward's® Chemistry Reduction of Vanadium Demonstration
Supplier: Avantor
470010-822EA 57.99 USD
470010-822
Ward's® Chemistry Reduction of Vanadium Demonstration
Educational Classroom Kits and Activities
The Ward’s Chemistry Teacher Demonstration is the perfect supplement to chemistry lectures.
  • Comes with enough chemicals to perform each demonstration five to seven times
  • Can be performed in just 10–30 minutes
  • Requires only basic labware that you provide
Watch the colors change as the force of agitation increases with this demonstration that shows a yellow solution turning green when it is gently shaken, blue when it is shaken slightly harder, and violet when the solution is shaken vigorously.