Chemistry Gram Stain Kit
Supplier: ALDON CORP SE
GG0221-KIT GG0221-PK/5
470010-662EA 39.5 USD
470010-662 470302-350
Chemistry Gram Stain Kit
Chemicals for Science Education
The Chemistry Gram Stain Kit makes gram straining easy and fast.
  • Shelf Life of 36 Months
  • Students Compare Microorganisms and Their Characteristics
  • No Chemical Mixing
  • Multiple Solutions Included
  • Fast and Simple Process
It lets you compare microorganisms and their characteristics and doesn't require chemical mixing.
Certifications: Note:This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Ordering information: Kit No. 470010-662 contains
30ml crystal violet
30ml iodine potassium iodide
30ml safranin O
100ml 95% ethanol
Kit No. 470302-350 contains
500ml crystal violet
500ml iodine potassium iodide
500ml safranin O
2 X 1000ml 95% ethanol