Cornstarch

Supplier: ALDON CORP SE

SS0950-500
470009-250PK 17.5 USD
470009-250
Cornstarch
Chemicals for Science Education

CAS Number: 9005-25-8
Formula Weight: (162.15)n
Formula: (C6H10O5)n
Density (g/mL): 1.45-1.6
Solubility: Hot Water
Synonyms: Maize Starch
Shelf Life (months): 12
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR