Dual Calibration Spring Scales
Supplier: Skywill Far East Industries
1712601
470008-100BX 1460.7 USD
470008-100
Dual Calibration Spring Scales
Balances and Scales
Economy Color-Coded Spring Scales
  • Provide direct readings for both force and mass measurements
  • Calibrated in both grams and Newtons
  • Can be fully zeroed
The body tube is constructed of clear plastic with color-coded plastic ends, and chrome-plated hooks and rings. The top of each spring is leveled for easy use on an inclined plane.
Caution: Caution:CHOKING HAZARD–These scales contain small parts and are not for children under three years.