Electric Gas Lighter
Supplier: Bel-Art
H15056-0000_SE
470008-096EA 87.15 USD
470008-096
Electric Gas Lighter
Heaters Gas Burners
A hazard-proof lighter, without an open flame, provides just what you need for safely lighting Bunsen burners, gas stoves, and propane or oxy-acetylene torches.
It eliminates the difficulties of striking flint and requires no batteries or fuel refills. Instead, the comfortable plastic pistol-grip has a push-button trigger that creates an electrical spark at 20 KV with each squeeze.