Somso® Animal Mitosis Set
Supplier: MARCUS SOMMER
ZOS 57
470008-026KT 1081.15 USD
470008-026
Somso® Animal Mitosis Set
Models Molecular Biology Models
Series Depicts Detail Of Nuclear Division
  • Each Stage Individually Mounted
  • 5 Year Manufacturers Warranty
From prophase, through metaphase and anaphase, to telophase, this set of models follows the process of mitosis. The separate forms, enlarged 2,000X, allow each stage to be studied one at a time, or collectively. Models 1–3 illustrate prophase; model 4, metaphase; models 5–7, anaphase; and model 8, telophase. Each model is mounted on its own stand. The set includes a manual describing the stages of mitosis. Size of each: 11 x 11 x 21 cm.