Grooved Director
Supplier: Dunrite Instruments
140670
470008-016BX 211.65 USD
470008-016
Grooved Director
Dissection Needles
This versatile instrument is several tools in one. Use the pointed end like a dissecting needle, and the grooved flat end can be used as a section lifter or to 'hold' an artery or nerve in the groove to make it easier to see and trace. Stainless steel.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.