Ward's® Chemistry of Fats Lab Activity
Supplier: Avantor
470007-970KT 107.29 USD
470007-970
Ward's® Chemistry of Fats Lab Activity
Educational Classroom Kits and Activities
Understand the Structure and Function of Fats
  • Understand the difference between saturated and unsaturated fats
  • Visually see how triglyceride synthesis occurs
As students construct models of glycerol and fatty acid sub-units and then put them together to form lipids, they will be able to visualize how triglyceride synthesis works. In the process, they will also learn the difference between saturated fats and unsaturated fats and how these two types of lipids can affect a healthy diet.
Includes:
Molecular Model Packets
Teacher Guide/Student Guide