Denoyer-Geppert® Giant Ear Model
Supplier: Denoyer-Geppert Science
0133-00
470007-846EA 519.75 USD
470007-846 470112-562
Denoyer-Geppert® Giant Ear Model
Models Human Biology Models
The fluid-filled canals of this model demonstrate the concept of equilibrium.
  • Model Enlarged Five Times
  • Removable Parts
  • Color-Coded Canals
  • Models Show How Ears Control Body's Equilibrium
Perfect for anatomy, biology, and speech therapy courses, the 5X-enlarged model presents all ear structures in magnificent detail. Its most unique feature, two semicircular canals filled with fluid, allow students to read the air bubble as if it were a level. The labyrinth and cochlea can be removed to display the vestibule, auditory nerve, and stapes; the cochlea is dissected to show the spiral membrane that aids hearing.
The three canals are color coded. The tympanum, malleus, and the incus are also removable.