Digital Multimeter with Transistor Test
Supplier: B & K Precision
2704C
470007-814EA 164.9 USD
470007-814
Digital Multimeter with Transistor Test
Electrical Meters Digital Multimeters
All of the functionality for basic Digital Multimeter applications including some high end features.
The 2000 count LCD can display resistance values up to 2 Gohm, capacitance to 20 uF or a transistors common-emitter gain value up to 1000. Value-packed features make this meter a must in every tool kit.