Stream Table Recirculating Pump
Supplier: Avantor
470007-780EA 199.99 USD
470007-780
Stream Table Recirculating Pump
Models Stream Models
Regulated, Constant Supply of Water
Much easier and less messy than adding water by hand, the 1/70 hp pump circulates up to 300 gallons per hour. It comes with a reservoir and mounting bracket. 115 VAC, 60 Hz. Reservoir size: 14"L x 12"W x 4"H.