Seawater Analysis Lab Activity
Supplier: Kemtec
4-102
470007-748KT 713 USD
470007-748
Seawater Analysis Lab Activity
Educational Materials Earth Science Educational Materials Natural Resources Learning Activities
Take an Ocean Voyage Without Leaving Your Classroom
Students analyze the chloride content of seawater by performing titrations using the precipiometric method. The included samples are from the Atlantic and Pacific Oceans, as well as from the Gulf of Mexico. Water from a local source can be analyzed as well. With the included materials, each titration can be repeated three times. It comes with enough materials for 12 setups, student data sheets, and instructional booklet.