Blood Typing Using Saliva Lab Activity
Supplier: Kemtec
3-214
470007-746KT 278.25 USD
470007-746
Blood Typing Using Saliva Lab Activity
Educational Materials Biology Educational Materials Physiology Learning Activities
This kit lets students determine their blood types without drawing a single drop of blood.
  • Material for 24 Students
  • Saliva Determines Antigens and Blood Type
  • No Blood Drawing Needed
  • Centrifuge Required
Students use their saliva to determine if they are among the 80% of the population who secrete antigens. Those who are then determine their ABO blood type.
The kit includes material for 24 students. A centrifuge is required but not included.
Ordering information: This item contains a coupon for perishable materials that must be redeemed from the manufacturer before use. Please allow sufficient time to account for redemption. These materials are shipped from the supplier via 2nd day delivery on Mondays, Tuesdays, or Wednesdays only and must be received promptly.