Boreal Science Chlorophyll Chromatogram/Wavelength Demonstration
Supplier: Avantor
470007-712KT 132.89 USD
470007-712
Boreal Science Chlorophyll Chromatogram/Wavelength Demonstration
Educational Classroom Kits and Activities
Demonstrate the presence of different chlorophyll compounds in plants using thin-layer chromatography.
Students will see the ways in which chromatograms and wavelength determination are used in chemical procedures. Includes developing solvent and concentrated chlorophyll extract sufficient for 100 demonstrations of a paper chromatogram and 100 demonstrations showing the wavelengths absorbed by the chlorophyll, and instructions.
Caution: Due to the nature of this item, it cannot be shipped by air under any circumstances. Please select ground shipping at time of checkout.