Somso® Comprehensive Eye Model
Supplier: MARCUS SOMMER
CS 5
470007-338EA 484.42 USD
470007-338
Somso® Comprehensive Eye Model
Models Human Biology Models
A highly detailed Eye Model.
  • Eyeball Is 4x Sized
  • Removable from the Stand for Easy Examination
  • Five-Year Manufacturer's Warranty
  • Exceptional Detail for Advanced Studies
The sclera is sectioned horizontally to show attached ocular muscles and vessels, and the choroid is medically sectioned to show the retina, ora serrata, and optic nerve. Enlarged four times, with six removable parts, the detailed model provides a great picture of the overall structure of the eye and the interrelationships of the individual components.
Removable parts include the two-part sclerotic membrane, vitreous humor, and lens.