Ward's® Sordaria Crossing-Over Flashcards
Supplier: PROFORMA ASSOCIATES
323377
470007-268KT 43.8 USD
470007-268
Ward's® Sordaria Crossing-Over Flashcards
Educational Materials Biology Educational Materials Biology Flashcards
Striking Images Capture Meiotic Recombination
  • Perfect companion to any crossing over lab activity
Determine the percent of crossing over that occurs in Sordaria asci by using the different photomicrograph shown on each card. The set is a perfect companion to any crossing over lab activity or when you are unable to prepare your own slides.