Angle of Insolation Kit
Supplier: Avantor
470007-262KT 283.89 USD
470007-262
Angle of Insolation Kit
Educational Classroom Kits and Activities
A Simple Alternative to Large Angle of Insolation Energy Labs
  • Convinient
  • Easy to Demonstrate
This conveniently sized classroom kit includes everything you need to perform the classic energy experiment without the hassle of extra lights, clamps, and ringstands. Developed by Steve Cronmiller, this versatile kit can also demonstrate the heating and cooling rate of land water and energy absorption when you add your own cans and substrates. Includes instructions.