Density Blocks II: Constant Mass
Supplier: Avantor
470007-252EA 61.49 USD
470007-252
Density Blocks II: Constant Mass
Educational Classroom Kits and Activities
This set of blocks, developed by Jodye Selco, is an excellent complementary set to Density Blocks I.
  • Grades 4 to 10
  • Set of eight
Students will observe that although each block has approximately the same mass, the size of each block is different because the density of each is different. The set includes one each of pine, oak, PVC, polypropylene, acrylic, aluminum, brass and steel. Includes a sturdy storage box and teacher’s guide with activities.