Ward's® Fermentation Kit
Supplier: Avantor
470007-198KT 41.59 USD
470007-198
Ward's® Fermentation Kit
Specimens Live Specimens Fungi - Living Material
Re-create the centuries-old process of food preservation and alcohol production
  • Ferment your own fruit or juice (not for consumption)
By treating grape or apple juice that you supply with a yeast culture, then monitoring for carbon dioxide evolution as the juice’s sugars are converted to alcohol, students learn about the process of fermentation.
This versatile kit also allows you to expand on the exercise by beginning with fresh grapes, or continuing to observe as the alcohol becomes vinegar.
It comes with instructions and enough materials for three demonstrations. Note: Includes perishable materials. Please specify delivery date.
Ordering information: This item contains a copupon for live or perishable materials. At checkout, please provide the date you wish the perishable components to arrive.
Delivery information: The kit will ship separately from the live component. Kits do not contain perishable components and generally ship via UPS ground.
Perishable components will ship via overnight or second day delivery.
Caution: While the organism in this activity is non-pathogenic, Ward’s recommends that you utilize proper aseptic technique when handling all microorganisms.
This kit is for educational use only. Do not consume any food or beverage product that has been fermented as a result of this kit.