Steel Ball Set
Supplier: Avantor
470007-130KT 12.35 USD
470007-130
Steel Ball Set
Educational Materials Physics Educational Materials Mechanics Learning Activities
Set of three undrilled balls.
One each 16, 19 and 25 mm diameters.
Un-drilled are suitable for collision apparatus and density/volume experiments. Balls available in sets or individually.