Determination of Chemical Formulas Student Activity
Supplier: Lab-Aids
85
470007-034EA 154.2 USD
470007-034
Determination of Chemical Formulas Student Activity
Educational Materials Chemistry Educational Materials Properties of Matter Learning Activities in Chemistry
Predict the Various Formulas of Compounds Formed by Their Reactions
  • Turn abstract theory into realism
  • Gain and understanding of percentage composition and molecular weight
  • Requires one 50 minute class period
Students predict the various formulas of compounds formed by their reactions. Strengthens lab procedure skills and observation abilities, and promotes an understanding of percentage composition and molecular weight.
Ordering information: Materials include Chemplates®, Lead nitrate solution, Sodium oxalate solution, Potassium iodide solution.