Seed Dispersal Riker Mount
Supplier: WHITE OWL ENTERPRISES
R322
470006-964EA 64.95 USD
470006-964
Seed Dispersal Riker Mount
Specimens Preserved Specimens Display Specimens
Display Examples of Four Different Dispersal Strategies
Bring the outdoors right inside your classroom to illustrate how seeds are dispersed. Twelve representative seeds are included and the descriptive overlay explains how these seeds are disseminated by wind, water, animals, and mechanical means. Size: 8" x 12" each.