LaMotte® Plant Tissue Macronutrient Test Kit
Supplier: LaMotte
5026-01
470006-954KT 173.9 USD
470006-954
LaMotte® Plant Tissue Macronutrient Test Kit
Educational Materials Biology Educational Materials Plant Learning Activities
What Soil Nutrients Do Your Plants Really Use?
  • Select your own plant tissue extracts
  • Convenient Carrying Case
  • Includes a Soil Handbook
Compare plants from home, the field, or a hydroponic greenhouse to determine the amount of nutrients they receive from the soil. Just a single plant tissue extract can be used for colorimetric tests of nitrogen, potassium, and phosphorus (fifty tests per factor), and all results can be quantified by trace, low, medium, or high concentrations.
The kit also comes with material for students to prepare their own plant tissue extracts. A soil handbook and all reagents and labware are supplied in a sturdy plastic case.