Force Table Pulley with Clamp
Supplier: Wards
PH0301A
470006-820EA 64.76 USD
470006-820
Force Table Pulley with Clamp
Educational Materials Physics Educational Materials Mechanics Learning Activities
This force table pulley includes a heavy-duty clamp and operates smoothly.
  • Pulley Fits CENCO Force Tables
  • Ideal for Composition of Forces
  • Pulley Also Works for Inclined Plane Experiments
The metal clamp attaches securely to tabletops from 10 to 28 mm thick.
The pulley wheel runs smoothly on ball bearings. It can be used for inclined plane experiments and is ideal for composition of forces