Dry Ice Maker
Supplier: Bel-Art
F38886-0000_SE
470006-806EA 418.95 USD
470006-806
Dry Ice Maker
Ice Makers
Produce dry ice in flake or snow form efficiently and economically. Just screw the brass nozzle to an LCO2 cylinder, then open the cylinder valve. The liquid carbon dioxide expands through the cone and into the bag to form dry ice.
  • Screws onto any LCO2 cylinder equiped with a dip tube
The LCO2 cylinder must have a dip tube.
Ordering information: Brass nozzle and collector bag only. Liquid carbon dioxide needs to be provided.