Dry Ice Machine
Supplier: Bel-Art
H38876-0000
470006-804EA 1395.3 USD
470006-804
Dry Ice Machine
Ice Makers
Create a 450 g Block of Dry Ice in less that two Minutes. This sturdy unit features a brass valve, chemical-resistant polyethylene chamber, a durable epoxy-coated steel frame with four rubber feet to protect counters, and a porous polyethylene diffusion plate to vent excess gas.
  • Fast and efficient
  • Includes nickel-plated copper connecting tube (2 m) with standard threaded couplings
Simply connect an LCO2 cylinder to the inlet on the control valve, then let the liquid carbon dioxide flow into the polyethylene chamber. A flip of the bar clamp releases the ice.
Ordering information: Contact your local gas supplier for CO2 gas.