Sta-Clear Lens Cleaning Station, Sellstrom
Supplier: SUREWERX USA
23470
470006-752EA 137.5 USD
470006-752
Sta-Clear Lens Cleaning Station, Sellstrom
Cleaning Agents Lens Cleaning Solutions
These water-activated cleansing tissues are safe for glass or plastic lenses and contain an anti-fogging agent.
  • Wipe away grime in seconds
The enamel-finished, heavy-gauge metal Sta-Clear cleaning station cabinet easily mounts on the wall and contains a plastic water bottle and pump, a dispenser with 1000 Sta-Clear tissues, and a tissue disposal compartment.
Delivery information: Cabinet is supplied complete with key.