Hanna Instruments Salinity Test Kit
Supplier: Avantor
470006-748KT 75 USD
470006-748
Hanna Instruments Salinity Test Kit
Environmental Test Kits
This chemical test kit that measures salinity by mercuric nitrate titration. This is supplied complete with all of the reagents and equipment necessary to perform approximately 110 tests.
  • Pre-made reagents for ease of use
  • All reagents marked with expiration date and lot number for traceability
  • Manual titration performed with diphenylcarbazone indicator
This is a titration-based chemical test kit that measures salinity within the 0.0 to 40.0 g/kg range. The HI3835 is supplied with all of the necessary reagents and equipment to perform the analysis. The test kit contains enough reagents for perform approximately 110 tests. Salinity is defined as the total solids in water after all carbonates have been converted to oxides, all bromide and iodide have been replaced by chloride, and all organic matter has been oxidized. The salinity value is in g/kg or ppt (parts per thousand). The monitoring of salinity is essential for industrial waste and seawater, as different species of plants and animals thrive varying salinity levels.
Ordering information: Replacement reagents need to be purchased seperatly.
Caution: This product can expose you to chemicals including mercury and mercury compounds, which are known to the State of California to cause cancer. (Propisition 65)