Somso® Human Development Model
Supplier: MARCUS SOMMER
MS 15/1
470006-640EA 1686.7 USD
470006-640
Somso® Human Development Model
Models Human Biology Models
Follow Development From Conception Through The End Of The First Month
  • First 30 Days Of Human Development In One Model
  • Easy To Store And Use
  • 5 Year Manufacturers Warranty
Thirteen individual models are mounted on a common base to allow your students to visualize the changes that occur during the first month of development. Models 1–9 are magnified 4,000X, and models 10–13 are magnified 4X. Each model is labeled directly on the base.