Fiber Optic Element
Supplier: Arcor Electronics
FOK-1
470006-588EA 64.45 USD
470006-588
Fiber Optic Element
Educational Materials Physics Educational Materials Optics Learning Activities
3' Fiber optic element, easily demonstrates the bending of light through the fiber optic wire. An inexpensive way to introduce students to fiber optics specifically the use of fiber optics for visual applications.
Teacher notes explain what fiber optic is and how it works and offers suggestions for activities. An economical way to prepare students for more advanced fiber optic studies/kits.