Electric Generator
Supplier: Jensen Manufacturing
15
470006-534EA 222.9 USD
470006-534
Electric Generator
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
This belt-driven generator may be used with our Steam Engine (63030-00 ). Operating at approximately 1500 rpm, the generator will produce enough electricity to light the included 3.8 volt bulb. The unit is mounted on a metal base (7.6×11.4 cm) and has a V-grooved pulley for the drive belt.