Light Combiner Demonstration
Supplier: Avantor
470006-468KT 65.99 USD
470006-468
Light Combiner Demonstration
Educational Classroom Kits and Activities
The Light Combiner is a simple yet dramatic way to demonstrate the additive color process.
  • Developed by Joe Rich
By placing the Light Combiner on an overhead projector and adjusting the focus, the primary light colors of red, green and blue are combined to produce the secondary light colors of yellow, cyan and magenta and white light. In addition to the instructions, this booklet includes a reproducible worksheet for students to record their observations.