Economy Digital and Analog Incubators
Supplier: Quincy Labs
10-140
470006-400EA 740.7 USD
470006-400
Economy Digital and Analog Incubators
Incubators Compact Benchtop Incubators
Economy gravity convection ovens feature a perforated heat shield for good temperature uniformity for quick run-up and recovery times.
  • Incubator dries, bakes, sterilizes, and evaporates
  • Maximum temperature: 62°C (145°F)
  • Scratch-resistant finish
  • Dual-range laboratory thermometer
  • Three-wire cord and plug
  • Aligned with GLOBE protocols
They also have heavy-steel, double-wall construction and a corrosive-resistant, aluminized steel interior.
The light gray hammer finish is scratch resistant. It has two adjustable chrome-plated wire shelves and supports.
Certifications: Constructed with UL and CSA-listed components.
Ordering information: The incubator includes a dual-range laboratory thermometer and a three-wire cord and plug. Supplied with an 18 month factory warranty.