Fire Piston Demonstration Apparatus
Supplier: American Scientific
1522-00
470006-372EA 30.95 USD
470006-372
Fire Piston Demonstration Apparatus
Educational Materials Physics Educational Materials Mechanics Learning Activities
Dramatically Demonstrate that Temperature Increases with Pressure
  • Includes instructions
  • Size: 8"H x 2 1/2" dia.
A piston tipped with three O-rings is pushed into a heavy glass tube to compress the air inside. Cotton at the bottom of a tube will ignite when the pressure in the tube is increased rapidly to show a temperature increase when a fixed mass of gas is compressed. You can demonstrate the need for oxygen to support combustion by showing that re-ignition will only occur if the piston is completely removed to allow a fresh supply of air. The apparatus can also be used to demonstrate cloud formation when the reduction in pressure lowers the temperature and immediately condenses the water vapor inside the cylinder.