Air Table with Air Supply
Supplier: The Science Source
12000
470006-324EA 1167.9 USD
470006-324
Air Table with Air Supply
Educational Materials Physics Educational Materials Mechanics Learning Activities
No more trying to chase down the air supply with this apparatus. Now you can buy an air table with an internal air supply for less than the cost of most tables alone.
  • Size: 60×91 cm
This unit combines sturdy construction and good blower performance for efficient, stable operation at a surprisingly low noise level. The smooth surface is finished in black, the best color for photo work.
Other features include adjustable tension bumper wire, threaded inserts for puck launcher, easy leveling, and a conveniently located on-off switch. Comes with a three-wire cord for operation on 120 VAC.