Fruit and Potato Clock
Supplier: VWR International
PT-CLK
470006-220EA 29.2 USD
470006-220
Fruit and Potato Clock
Educational Materials Chemistry Educational Materials Electrochemistry Learning Activities
This LCD clock keeps time using potatoes, soda, or plants.
  • Length: 8½"
  • Width: 3½"
  • Height: 2¾"
  • Demonstration of an Electrochemical Cell
  • Instructions Included
It's an ideal demonstration of an electrochemical cell. All you need to do is attach the included zinc and copper bimetallic probes and watch the resettable hours, minutes, seconds, and date light up.
The clock includes instructions and measures 2¾ x 3½ x 8½".