Hand-Driven Illuminated Planetarium
Supplier: Science First
653-3010
470006-100EA 692 USD
470006-100
Hand-Driven Illuminated Planetarium
Models Astronomy Models
Study different concepts utilizing sunlight or darkness.
  • Demonstrates a variety of phenomena
  • Sun can proviide illumination
  • Includes a study guide
Use the gear-and-chain-driven planetarium in daylight to study seasons and the tides, or darken the room and the illuminated sun allows investigation of moon phases, eclipses, midnight sun, twilight, dawn, length of day, and more. A study guide with activities is included. 110 VAC with a 5' cord. Size: 20"L x 14"W.