Fischer Technical Universal Vacuum Plate
Supplier: Fischer Technical
GA-001
470006-052EA 440.6 USD
470006-052
Fischer Technical Universal Vacuum Plate
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
This nickel coated plate uses a rubber pad to achieve air tight contact with a bell jar. It features a stopcock and center hole with safety screw plug and a rubber gasket.
To create the vacuum, air is removed via a barbed hose connector that accommodates tubing of 6.4 mm (¼") int.Ø. Measures diameter 27 cm with a 6 mm diameter center hole.