Specimen Tongs
Supplier: Dunrite Instruments
6343500
470005-898EA 4.7 USD
470005-898
Specimen Tongs
Tongs
Ideal for handling preserved specimens, the wide clamping jaws are flattened and have serrations to prevent slipping.
The jaw spread is 2 inches. The tongs are constructed of nickel-plated steel and have an overall length of 8 inches.