Wax Marking Pencils
Supplier: PRECISION MOULDED POLYMERS LIMITED
M80851
470005-680EA 1.49 USD
470005-680 470005-682
Wax Marking Pencils
Pens Lab Markers
These excellent quality wax pencils are ideal for writing directly on glass, porcelain, metal, and ceramic ware. They may also be used on overhead transparencies. Reqires a sharpener, not included.