Spherical Mirrors
Supplier: Wards
PH0526E MCV302 PH0520C PH0520D PH0520E PH0526C PH0526D MCC302 MCC703 MCC304 MCC306 MCC310 MCV304 MCV306 MCV310 PH0520F
470013-316EA 6.84 USD
470013-316 470005-646 470013-304 470013-306 470013-308 470013-312 470013-314 470014-718 470016-996 470111-298 470111-300 470111-302 470111-304 470111-306 470111-308 470013-310
Spherical Mirrors
Mirrors
These wide range of mirrors are perfect for investigating real and virtual images.
  • Variety of Sizes and Focal Lengths
  • Circular
  • Polished Glass Construction
  • Durable
  • Back-Surface Silvered
The circular mirrors are available in diameters of 38 mm, 50 mm, and 75 mm.
They're back-surface silvered, made of polished glass, and painted for durability.