Handheld Discharge Electrode
Supplier: Science First
N-122
470005-530EA 158.45 USD
470005-530
Handheld Discharge Electrode
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
The Handheld Discharge Electrodeeasily and safely discharges an electrostatic generator.
  • Polished Aluminum Sphere
  • Insulated Lucite Rod
  • Flexible Wire Lead
  • Generator Attaches to Wire Lead
  • Electrode Discharges an Electrostatic Generator
A polished aluminum sphere, 10.5 cm in diameter, is mounted on a 40 cm insulated Lucite rod.
A flexible wire lead has a banana plug at one end and a clip on the other, so it can attach to the base of the generator.