Screw Lock Scalpel
Supplier: Dunrite Instruments
140705
470005-456EA 7.55 USD
470005-456
Screw Lock Scalpel
Scalpels and Scalpel Assemblies Scalpels
A versatile, inexpensive scalpel with a safety lock to hold the blade securely, yet allow for simple, safe changes.
The handle is fluted for a better grip, with a blade best suited for heavy dissections. The blade is held in place using a pin and screw spaced for blades fitting a number 4 scalpel.