Hoffman Screw-Compressor Clamps for Flexible Tubing
Supplier: United Scientific Supplies
HSCOP2
470005-380EA 5.85 USD
470005-380
Hoffman Screw-Compressor Clamps for Flexible Tubing
Clamps General Purpose Clamps
Easily clamp tubes up to 1" in diameter.
  • Zinc-plated brass
Can use as a closed clamp or open.