Symmetrical Round Jaws Double Buret Clamps
Supplier: GSC International
4-BCDR86
470005-374EA 17.7 USD
470005-374
Symmetrical Round Jaws Double Buret Clamps
Clamps General Purpose Clamps
Use the two sets of symmetrical round jaws for simultaneously holding two round objects, each up to 35 mm in diameter. Each of the rubber-coated clamps rotates 360° and may be locked at any angle. The unit attaches to any standard support rod up to 13 mm in diameter.